missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacillus Unsaturated glucuronyl hydrolase Protein

A DNA sequence encoding the Bacillus sp. (strain GL1) Unsaturated glucuronyl hydrolase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8079-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002541

  • 865.00 EUR / 100µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Unsaturated glucuronyl hydrolase Protein
Research Use Only
ugl
Recombinant Protein
E. coli
MWQQAIGDAL GITARNLKKF GDRFPHVSDG SNKYVLNDNT DWTDGFWSGI LWLCYEYTGD EQYREGAVRT VASFRERLDR FENLDHHDIG FLYSLSAKAQ WIVEKDESAR KLALDAADVL MRRWRADAGI IQAWGPKGDP ENGGRIIIDC LLNLPLLLWA GEQTGDPEYR RVAEAHALKS RRFLVRGDDS SYHTFYFDPE NGNAIRGGTH QGNTDGSTWT RGQAWGIYGF ALNSRYLGNA DLLETAKRMA RHFLARVPED GVVYWDFEVP QEPSSYRDSS ASAITACGLL EIASQLDESD PERQRFIDAA KTTVTALRDG YAERDDGEAE GFIRRGSYHV RGGISPDDYT IWGDYYYLEA LLRLERGVTG YWYERGR
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
Bacillus
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers