missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacillus Levanase Protein

A DNA sequence encoding the Bacillus sp. (strain L7) Levanase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7478-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028950

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Levanase Protein
Research Use Only
Partial (see sequence information for more details).
Bacillus
His
Tris-based buffer, 50% glycerol
1 mg
Not determined.
Recombinant Protein
Yeast
LPWNDLGHVW SGSAVADTTN ASGLFGSSGG KGLIAYYTSY NPDRHNGNQK IGLAYSTDRG RTWKYSEEHP VVIENPGKTG EDPGGWDFRD PKVVRDEANN RWVMVVSGGD HIRLFTSTNL LNWTLTDQF
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers