missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant B. abortus Malate dehydrogenase Protein
A DNA sequence encoding the Brucella abortus (strain S19) Malate dehydrogenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6351-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
WP_002964995.1 | |
200 μg | |
Not determined. | |
Full Length | |
B. abortus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Malate dehydrogenase Protein | |
Research Use Only | |
mdh | |
Recombinant Protein | |
E. coli | |
MARNKIALIG SGMIGGTLAH LAGLKELGDV VLFDIAEGTP QGKGLDIAES SPVDGFDAKF TGANDYAAIE GADVVIVTAG VPRKPGMSRD DLLGINLKVM EQVGAGIKKY APEAFVICIT NPLDAMVWAL QKFSGLPAHK VVGMAGVLDS ARFRYFLSEE FNVSVEDVTV FVLGGHGDSM VPLARYSTVA GIPLPDLVKM GWTSQDKLDK IIQRTRDGGA EIVGLLKTGS AFYAPAASAI QMAESYLKDK KRVLPVAAQL SGQYGVKDMY VGVPTVIGAN GVERIIEIDL DKDEKAQFDK SVASVAGLCE ACIGIAPSLK | |
Greater than 88.5% as determined by SDS-PAGE. |