Learn More
Invitrogen™ Rat LMAN1 (aa 159-173) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP277
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Rat ERGIC-53 (residues 159-173) Synthetic Peptide, PEP-277, from Invitrogen.
PEP-277 is a 15 amino acid synthetic peptide whose sequence corresponds to residues 159-173 of rat ERGIC-53. The sequence of this peptide is (amino to carboxy terminus): F (159)-D-S-F-D-N-D-G-K-K-N-N-P-A-I (173).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, ERGIC-53, catalog # PA1-074. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
116666 | |
purified | |
Rat LMAN1 (aa 159-173) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Rat | |
Q62902 | |
0.35 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Lman1 | |
No preservative | |
35 μg | |
F(159) - D - S - F - D - N - D - G - K - K - N - N - P - A - I(173) |