missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IgG2b Kappa Light Chain Isotype Control (149/10H5), DyLight 488, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG2b Kappa Light Chain Monoclonal specifically detects IgG2b Kappa Light Chain in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | IgG2b Kappa Light Chain |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 149/10H5 |
| Conjugate | DyLight 488 |
| Dilution | Flow Cytometry 1:10 - 1:1000 |
| Formulation | 50 mM Sodium Borate. with 0.05% Sodium Azide |
| Host Species | Rat |
| Quantity | 500 Tests |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?