missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cd86, Rat, PE, Clone: GL1, Abnova™
Rat monoclonal antibody raised against native Cd86.
Brand: Abnova MAB5753.100ug
Additional Details : Weight : 0.00010kg
Specifications
Cd86 | |
Monoclonal | |
PE | |
Flow Cytometry (0.1 ug/106 cells) The optimal working dilution should be determined by the end user. | |
Cd86 | |
Cd86 | |
Native purified Cd86 from LPS-activated mouse B cells. | |
RUO | |
12524 | |
Mouse | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
GL1 | |
Rat monoclonal antibody raised against native Cd86. | |
In PBS (0.09% sodium azide) | |
B7/B7-2/B7.2/B70/CLS1/Cd28l2/ETC-1/Ly-58/Ly58/MB7/MB7-2/TS/A-2 | |
Rat | |
100 μg | |
Primary | |
Mouse CD86, the B7-2 co-stimulatory molecule and a ligand for CD28 and CD152/CTLA-4. | |
Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a κ |