missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASA3 Monoclonal Antibody (1F11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA521351
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: DGDRETERIY SLFNLYMSKL EKMQEACGSK SVYDGPEQEE YSTFVIDDPQ ETYKTLKQVI AGVGALEQEH AQYKRDKFKK TKYGSQEHPI GDKSFQNYIR QQSETSTHSI
RASA3 Monoclonal antibody specifically detects RASA3 in Human samples. It is validated for ELISA, Western Blot.Specifications
RASA3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q14644 | |
RASA3 | |
RASA3 (AAH47242, 725 a.a. ∽ 834 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
1F11 | |
Unconjugated | |
RASA3 | |
AI326412; C86362; E130011G04; GAP1(IP4BP); GAP1-InsP4 BP; GAP1IP4BP; GAPIII; GAPIII activator 3; GTPase activating protein III; GTPase-activating protein 1 family, inositol 1,3,4,5-tetrakisphosphate-binding protein; ins P4-binding protein; Ins(1,3,4,5)P4-binding protein; ras GTPase-activating protein 3; Ras GTPase-activating protein III; RAS p21 protein activator 3; Rasa3; RP11-245B11.3; R-Ras gap; R-Ras GTP activating protein; R-ras GTPase activating protein; R-ras RTPase activating protein-3; scat | |
Mouse | |
Protein A | |
RUO | |
22821 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |