Learn More
Tenak™ Classic Rack for Chest Freezers, with Locking Rod, 32 mm Box height
Classic Rack is suitable for use with all chest freezer models.
Brand: Tenak™ TE21350
Additional Details : Weight : 1.25000kg
Description
- Comes with a locking rod to secure the cryoboxes
- Measure the inside height of you freezer when choosing the right rack
- Constructed of stainless steel
Specifications
140 x 140 x 288 mm | |
Chest Frseezers | |
Locking rod |
Stainless Steel | |
Boxes (8) | |
32 mm |