missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slc14a2, Rabbit, R-PE, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant Rat Slc14a2.
Brand: Abnova PAB28836.100ug
Additional Details : Weight : 0.00010kg
Specifications
Slc14a2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant Rat Slc14a2. | |
PBS, 50% glycerol and 0.09% sodium azide | |
Slc14a2T/UrT1-C/UrT1-D/testSymbol | |
Rabbit | |
Affinity purification | |
RUO | |
54302 | |
Mouse, Rat | |
Liquid |
Immunofluorescence, Western Blot | |
R-PE | |
Immunofluorescence Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Slc14a2 | |
Slc14a2 | |
Recombinant protein corresponding to Slc14a2 C-terminus (amino acids 911-929). | |
100 μg | |
Primary | |
1 ug/mL of PAB28836 was sufficient for detection of Slc14a2, ∼97 and 127kDa, in 20ug of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody. | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only