missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD33 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CD33.
Brand: Abnova PAB3632.400uL
Additional Details : Weight : 0.02000kg
Specifications
CD33 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CD33. | |
In PBS (0.09% sodium azide) | |
NP_001763;P20138 | |
CD33 | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human CD33. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10-50) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
CD33 | |
FLJ00391/SIGLEC-3/SIGLEC3/p67 | |
Rabbit | |
Protein G purification | |
RUO | |
945 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only