missing translation for 'onlineSavingsMsg'
Learn More

Quinoxaline, 98+%, Thermo Scientific Chemicals

Catalog Number p-3758740
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
91-19-0
C8H6N2
130.15
MFCD00006719
XSCHRSMBECNVNS-UHFFFAOYSA-N
benzoparadiazine, 1,4-benzodiazine, phenopiazine, phenpiazine, quinazine, benzopyrazine, 1,4-diazanaphthalene, benzo a pyrazine, 1,4-naphthyridine, chinoxalin
7045
CHEBI:36616
quinoxaline
C1=CC=C2C(=C1)N=CC=N2
91-19-0
C8H6N2
130.15
MFCD00006719
XSCHRSMBECNVNS-UHFFFAOYSA-N
benzoparadiazine, 1,4-benzodiazine, phenopiazine, phenpiazine, quinazine, benzopyrazine, 1,4-diazanaphthalene, benzo a pyrazine, 1,4-naphthyridine, chinoxalin
7045
CHEBI:36616
quinoxaline
C1=CC=C2C(=C1)N=CC=N2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 29°C to 34°C
Color Yellow to Brown
Boiling Point 220°C to 223°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 23, 176
Merck Index 15, 8195
Solubility Information Solubility in water: soluble. Other solubilities: miscible in alcohol,ether,benzene,acetone
Formula Weight 130.15
Percent Purity 98+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Quinoxaline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'