missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinine hydrochloride, 100.8%, >99%, MP Biomedicals™
Click to view available options
Quantity:
25 g
50 g
Specifications
Color | White |
Assay Percent Range | 1.008 |
Quantity | 25 g |
Formula Weight | 360.9 |
Percent Purity | >99% |
Physical Form | Powder |
Chemical Name or Material | Quinine hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'