Learn More
Quickfit™ Female Spherical Ball Joint
For use with male ball joints MS:
Brand: Quickfit FS41UB
Additional Details : Weight : 0.10000kg
Description
For use with male ball joints MS:
- Female cup joints precision manufactured to the highest standards
- Spherical joints are manufactured with an approximate shank length of 100mm
Specifications
1/Pk. | |
Spherical Ball Joint | |
Shank Dia.: 28mm |
Bore: 27mm, Length: 100mm | |
S41 | |
2.8mm |