Learn More
Quickfit™ Female Spherical Ball Joint
For use with male ball joints MS:
Brand: Quickfit FS35UB
35.25 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.07000kg
Description
For use with male ball joints MS:
- Female cup joints precision manufactured to the highest standards
- Spherical joints are manufactured with an approximate shank length of 100mm
Specifications
Female Spherical Ball Joint | |
S35 | |
0.1in. | |
2.5mm |