Learn More
QuickFit™ Borosilicate Glass Test Tubes with Ground Socket
Click to view available options
Diameter (Metric):
14 mm
17 mm
19 mm
23 mm
29 mm
41 mm
Capacity (Metric):
5 mL
10 mL
11 mL
15 mL
19 mL
31 mL
54 mL
75 mL
195 mL
Unit Size:
Each
Pack of 5
Description
Manufactured from borosilicate glass for excellent chemical and thermal shock resistance. Quickfit™ Borosilicate Glass Test Tubes with Ground Socket are ideal for mixing and heating chemicals in a lab.
- Presicion-ground socket ensures exact fit for stopper
- Test tube sizes correspond with Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stoppers
- Order tubes and stoppers separately
Specifications
Specifications
| Design Type | Ground Socket |
| Description | Stopper Size: 19/26 |
| Capacity (Metric) | 31 mL |
| Length (Metric) | 150 mm |
| Material | Borosilicate Glass |
| For Use With | Quickfit Hollow Stopper, Fisher Scientific product code 12568855 |
| Quantity | 5 Pack |
| Diameter (Metric) | 23 mm |
| Product Type | Test Tube |