Learn More
Quickfit™ Allihn Condenser with Ground Cone
Ideal for reflux applications.
Brand:  Quickfit CX7/06/SC
209.00 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.24000kg
 View more versions of this product
Product Code. 11361845
  • 220.00 EUR / Each
Estimated Shipment: 22-07-2024
Description
Ideal for reflux applications.
  • Inner bulb sections provide increased cooling surface
  • With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
Specifications
Specifications
40/38
Allihn Condenser
Borosilicate Glass
Ideal for reflux work
365 mm
365 mm