Learn More
Miele™ Quarter Insert
Brand: Miele™ 6907650
Additional Details : Weight : 47.25000kg
Specifications
Insert, Quarter | |
12 x 165mm Test tubes. | |
320 mm | |
320 mm |
E 105/1 | |
A 13 lid | |
192 mm | |
1 Each |