Learn More
Miele™ Quarter Insert
Brand: Miele™ 6907640
Additional Details : Weight : 50.00000kg
Specifications
Insert, Quarter | |
Test Tubes up to 12 x 105mm | |
320 mm | |
200 mm |
E 104/1 | |
A 13 lid | |
132 mm | |
1 Each |