Learn More
Sartorius Quantitative Filter Papers
These Quantitative Papers are used for quantitative and gravimetric analyses as well as for pressure or vacuum filtration.
35.50 EUR - 59.60 EUR
Specifications
Format | Disc |
---|---|
Sterility | Non-sterile |
No. per Pack | 100 Pk. |
Fit for Purpose | Certified Quality |
Product Code | Brand | Diameter (Metric) | Particle Retention | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Particle Retention | Price | Quantity & Availability | ||||
15912860
|
Sartorius
FT-3-104-100 |
100 mm (3.9 in.) | 2 to 3 μm |
35.50 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
15942850
|
Sartorius
FT-3-103-100 |
100 mm (3.9 in.) | 3 to 5 μm |
35.50 EUR
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
||||
15932860
|
Sartorius
FT-3-105-100 |
100 mm (3.9 in.) | 5 to 8 μm |
35.50 EUR
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
||||
15962840
|
Sartorius
FT-3-101-142 |
142 mm (5.6 in.) | 12 to 15 μm |
59.60 EUR
Pack of 100 |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
15932880
|
Sartorius
FT-3-112-142 |
142 mm (5.6 in.) | 8 to 12 μm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Made of 100% cotton linters
- Ash-free (ash content ≤0.01% according to DIN 54370)
- Wet-strengthened
- Color-coded box for easy selection
- 100 discs per pack
Specifications
Disc | |
Non-sterile | |
100 Pk. | |
Certified Quality |