missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Grade 389 F Quantitative Filter Paper
Used for quantitative and gravimetric analyses as well as for pressure or vacuum filtration.
Brand: Sartorius™ FT-3-112-185
Additional Details : Weight : 0.01000kg
Description
- Made of 100% cotton linters
- Ash-free (ash content ≤0.01% according to DIN 540370)
- Wet-strengthened
- Color-coded box for easy selection
Specifications
0.19mm | |
100Pack | |
Use to determining the fate content in natural raw materials |
Quantitative Filter Paper | |
185 mm |