Learn More
Socorex™ Qualitix™ Micropipette Tips, 1000μL, blue
Superior, autoclavable micropipette tips cover a range of sizes and fit a broad array of micropipette brands. Socorex™ Qualitix™ Micropipette Tips, 1000μL also come in a variety of shapes to facilitate fluid transfer.
Brand: Socorex™ 3190.1000RS
Additional Details : Weight : 0.41000kg
Description
- Cover a range of sizes.
- Autoclavable.
- Optimal tightness fitting on Socorex™ pipettes and broad compatibility with other brands.
- Various tip versions available.
Specifications
1000 μL | |
Non-Filtered | |
Blue | |
Regular Tip, Bevel Point | |
6/PK, 8PK/CS | |
Socorex™ |
1000μL | |
Sterile | |
Autoclavable | |
96/Rack | |
Single-use | |
Qualitix |