missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Qualitative/Technical Papers, Smooth, Sheets, Grade 3 hw
Suitable for analytical and technical analyses and featuring medium fast filtering
Brand: Sartorius Stedim Biotech™ FT-2-303-100100
Additional Details : Weight : 0.06500kg
Description
- Generally used for analytical and technical purposes.
- Smooth finish.
- Sheet format.
- Refined pulp and linters.
- Non-sterile, bulk packed.
- Suitable for clarification, analytical sample preparation, particle analysis.
Specifications
65 g/m2 | |
Cotton Linters, Refined Pulp | |
3 hw | |
0.14 mm |
Sheet | |
20 sec. | |
100/Pk. | |
100 x 100 mm |