Learn More
Alfa Aesar™ QuadraPure™ DET, 450-650 micron, metal scavenger
Metal Scavenger: Pd in acidic media and with phosphines, Co, Fe, Ni, Rh
Brand:  Alfa Aesar™ 046083.14
Additional Details : Weight : 0.02500kg
Quantity 25g
 View more versions of this product
Product Code. 15487145
  • 339.00 EUR / 25g
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Chemical Identifiers
QuadraPure™ DET metal scavenger
MFCD08705362
QuadraPure™ Bis(ethyl mercaptan)
Specifications
QuadraPure™ DET metal scavenger
MFCD08705362
450 to 650 Micron
25g
Beads
QuadraPure™ Bis(ethyl mercaptan)
White to beige