missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ QB Series Safety Cover

Catalog Number p-8014637
This item is not returnable. View return policy
This item is not returnable. View return policy

Made from tough clear acrylic for maximum visibility. Grant Instruments™ QB Series Safety Covers prevent accidental touching of a hot block or contamination of samples from splashes.

  • For use with QB Series Dry Block Heaters
  • Acrylic construction provides increased visibility
  • Prevents burning from accidentally touching the hot block
  • Clearance height of 85mm
Color Clear
Material Acrylic
Type Accessory, Safety Cover
For Use With (Application) For maximum visibility whilst preventing accidental touching of a hot block or contamination of samples from splashes
Height (Metric) 85 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'