Learn More
PYREX™ Class A Volumetric Flasks with Glass Standard Taper Stopper
Machine-blown body
Brand: PYREX™ 34505-34
Additional Details : Weight : 0.80000kg
Description
The strength of these flasks has been increased appreciably through machine-blown bodies to which are sealed heavy-tubing necks tooled for standard taper glass stoppers. The graduation line is sharp and permanent; large white block letters make the inscription easy to read. The 1 mL and 2 mL sizes are test tube shaped. Sizes 3L through 6L have pennyhead standard taper stoppers.
For stopper only, see Cat. Nos. 7650 and 7661.
Specifications
Flat | |
Pennyhead | |
Glass | |
Borosilicate Glass | |
Long | |
1/Pk. |
38 | |
20.5 cm | |
4 L | |
48 cm | |
1/Cs. | |
±1 mL |