Learn More
Pyrex™ Class A Glass Cylinder with Polyethylene Stopper
Manufactured from chemically resistant Pyrex™ borosilicate glass.
Brand: Pyrex 3212/04
57.62 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
- Complies with ISO 4788
- Printed with a batch code for downloadable batch certificate
- Manufactured from chemically resistant Pyrex borosilicate glass
- Graduations in white enamel for improved visibility with dark colored solutions
- Supplied with polyethylene stopper
- Robust hexagonal glass foot
Specifications
Graduated Cylinder | |
Glass | |
Class A | |
159 mm | |
10/19 |
Hexagonal | |
ISO 4788 | |
10 mL | |
Polyethylene Stopper | |
Polyethylene |