Learn More
Cole-Parmer™ PVDF Manifold, 4 port
Expand your flow and line capacity with these manifolds. Choose the number of ports and connections to suit your needs.
Brand:  Cole-Parmer™ 31523-41
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11785278
Quantity Price
1 198.00 EUR / Each
6 184.64 EUR / Each
Estimated Shipment
16-06-2021
Add to basket
Description
Description
  • Constructed of durable PVDF.
  • Ideal for applications requiring excellent chemical compatibly.
  • Available in a variety of configurations
Specifications
Specifications
230°F
110 psi
4.75 in.
Manifold
-54°C
1/2 in. NPT(F)
3.81 cm
110°C
PVDF
12.07 cm
-65°F
1/2 in. x 3/8 in.
3/8 in. NPT(F)
3.81 cm