Learn More
Ark-Plas Products™ PVC Tubing
Made of polyvinyl chloride. Ark-Plas Products™ Red PVC Hoses are ideal for use with connecting hoses of various diameters. Good flexibility and chemical resistance make this tubing ideal for use with apparatus assemblies.
Brand:  Ark-Plas Products™ TCX28-FC5
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11767709
  • 113.04 EUR / Each
Estimated Shipment
26-04-2021
Add to basket
Description
Description
  • Chemical and corrosion resistance
  • Abrasion and wear resistance
  • Rubber-like flexibility
Specifications
Specifications
Green
PVC
100 m
4.7625 mm
6.35 mm