Learn More
Ismatec™ Pump Head for REGLO-CPF Digital Pump Drive
Ceramic piston and cylinder pump head for the REGLO-CPF Digital Pump Drive.
Brand:  Ismatec™
Additional Details : Weight : 0.88000kg
Product Code. 15715103
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • For use with REGLO-CPF Digital Pump Drive
  • Ceramic Piston
  • Ceramic Cylinder
Specifications
Specifications
Stroke volume: 2.5-25 μL/revolution, Revolutions/minute: 40-1800, Flow ports: Kynar, UNF 1/4 in.-28 (female).
100°C
100 psi
0.1-45 mL/min.
Lip seals: Rulon AR
Kynar (PVDF)