Learn More
Abnova™ PTP4A3/CEN8q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0170.200uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16023265
  • 2060.00 EUR / 200µL
Estimated Shipment: 31-01-2023
to see stock.
Includes: DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Specifications
Liquid
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
FISH-Ce
Store at 4°C in the dark.
200 μL
11156
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe