missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGER4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534122
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey, Marmoset (100%) Guinea pig (88%) Elephant, Bat, Rabbit, Horse (82%).
The protein encoded by this gene is a member of the G-protein coupled receptor family. This protein is one of four receptors identified for prostaglandin E2. This receptor can activate T-cell factor signaling. It has been shown to mediate PGE2 induced expression of early growth response 1, regulate the level and stability of cyclooxygenase-2 mRNA, and lead to the phosphorylation of glycogen synthase kinase-3. Knockout studies in mice suggest that this receptor may be involved in the neonatal adaptation of circulatory system, osteoporosis, as well as initiation of skin immune responses.Specifications
PTGER4 | |
Polyclonal | |
Unconjugated | |
PTGER4 | |
EP1; EP4; EP4R; MGC126583; PGE receptor EP4 subtype; PGE receptor, EP4 subtype; PGE2 receptor EP4 subtype; prostaglandin E receptor 4; prostaglandin E receptor 4 (subtype EP4); prostaglandin E2 receptor EP4 subtype; prostaglandin E2 receptor type 4; prostanoid EP4 receptor; Ptger; PTGER2; Ptger4; Ptgerep4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5734 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.9 mg/mL | |
PBS with 0.1% sodium azide | |
P35408 | |
PTGER4 | |
Synthetic 17 amino acid peptide from 3rd extracellular domain of human PTGER4 / EP4. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |