Learn More
JG Finneran Associates™ PTFE/Rubber Seals
126.00 EUR - 164.00 EUR
Specifications
Material (Liner) Rubber
Color Red
Quantity 1000 Pk.
View More Specs
Products 2
Product Code Brand Price Quantity & Availability  
Product Code Brand Price Quantity & Availability  
10015580
View Documents Promotion Details
JG Finneran Associates™
604040-08
Size: 8mm
126.00 EUR
Pack of 1000
View alternative products
Save Up to 
»
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
12422920
View Documents Promotion Details
JG Finneran Associates™
604040-13
Size: 13mm
164.00 EUR
Pack of 1000
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
Specifications
Rubber
Red
1000 Pk.