Learn More
Hanna Instruments™ PTFE Membrane Dissolved Oxygen Probe
Brand:  Hanna Instruments™ HI76407/10F
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11517829
  • 822.00 EUR / Each
Estimated Shipment: 10-06-2024
Specifications
10 m
Standard DO
For both laboratory and field applications.
Thermistor temperature sensor