Learn More
Cole-Parmer™ PTFE Manifold, 4 outlets
Organize your fluid paths with these thermoplastic manifolds
Brand: Cole-Parmer™ 31521-11
Additional Details : Weight : 0.80000kg
Description
- PTFE Manifold.
- Chemically-inert PTFE handles the most aggressive chemical solutions.
- Available in a variety of configurations to suit a wide range of applications.
Specifications
PTFE Manifold, 4 Outlets | |
4.75 in. | |
-200°F | |
1/2 in. NPT(F) | |
3.81 cm | |
3.81 cm | |
120 psi | |
150°C |
PTFE | |
12.07 cm | |
-129°C | |
3/8 in. NPT(F) | |
1.5 in. | |
1.5 in. | |
300°F |