Learn More
BRAND™ PTFE Coated Steel Core Stirring Rods
Unique high-density PTFE coated polyethylene stirring rods with a steel core with rounds ends.
Brand:  BRAND™ 136320
17.81 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10161871
  • 18.75 EUR / Each
Estimated Shipment: 03-06-2024
Description
  • PTFE coated steel core stirring rods with round end
  • High-density polyethylene (with steel core)
  • Can be bent as required
  • The shape is permanently retained
Specifications
Specifications
6 mm
PTFE Coated Steel Core
1
250 mm