missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ PTFE-Coated Aluminum Block for RapidVap™ N2/48

PTFE-Coated Aluminum Blocks for use with RapidVap N2/48 Evaporation Systems.

Brand:  Labconco™ 7482300

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11736649

  • 1974.00 EUR / Each


Estimated Shipment
17-03-2021
Add to basket

Description

Description

  • All RapidVap N2/48 Evaporation Systems require a Block (not included)
  • Glassware is required (not included)
  • Custom blocks to fit special glassware such as VOA vials are available upon request
  • All RapidVap N2/48 Evaporation Systems require a Block (not included)
  • PTFE-coated aluminum for corrosion resistance
  • Holds 48 tubes up to 23mL
Specifications

Specifications

Up to 23 mL
RapidVap N2/48 Evaporation Systems
Accessory, Block
PTFE-coated Aluminum
48 tubes
Videos
SDS
Documents