Learn More
VACUUBRAND™ PTFE Antistatic Tubing
Offering superior chemical resistance in the laboratory.
Brand:  VACUUBRAND™ 22614366
Additional Details : Weight : 0.01000kg
Product Code. 11900642
  • 995.00 EUR / Each
Estimated Shipment: 25-06-2024
Description
  • 16mm diameter
  • 2500mm length
Specifications
Specifications
Tubing
PTFE
250 cm
Antistatic
16 mm