missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTEN Mouse anti-Human, Unconjugated, Clone: 5C10B6, Proteintech
Mouse Monoclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | PTEN |
---|---|
Clone | 5C10B6 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827385
|
Proteintech
60300-1-IG-20UL |
20 μL |
140.00 EUR
20µL |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16817385
|
Proteintech
60300-1-IG-150UL |
150 μL |
399.00 EUR
150µL |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The PTEN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PTEN. This antibody recognizes human, mouse antigen. The PTEN antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
PTEN | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
5728 | |
Primary |
5C10B6 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Mouse | |
IgG2a |