missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Proteinase K

From Tritirachium album

140.00 EUR - 310.00 EUR

Specifications

CAS 39450-01-6
pH Range 6.2 to 6.8 (20°C, 10 g/L in H2O)
Storage Requirements Store at 2°C to 8°C
Synonym Endopeptidase K, Proteinase K from Tritirachium album, Tritirachium alkaline proteinase
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10274070
View Documents
Merck Millipore
1245680100
100 mg 140.00 EUR
100mg
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment: 02-08-2021
Add to basket
 
10036570
View Documents
Merck Millipore
1245680500
500 mg 310.00 EUR
500mg
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment: 02-08-2021
Add to basket
 
Description

Description

  • Chromatographically purified lyophilized 30 m Anson-U/mg
  • For biochemistry and molecular biology EC 3.4.21.14
  • Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA.
  • Removes endotoxins that bind to cationic proteins such as lysozyme and ribonuclease A.
  • Reported useful for the isolation of hepatic, yeast, and mung bean mitochondria
  • Determination of enzyme localization on membranes
  • Treatment of paraffin embedded tissue sections to expose antigen binding sites for antibody labeling.
  • Digestion of proteins from brain tissue samples for prions in Transmissible
  • Spongiform Encephalopathies (TSE) research.
  • Proteinase K is a stable and highly reactive serine protease.
  • Evidence from crystal and molecular structure studies indicates the enzyme belongs to the subtilisin family with an active-site catalytic triad (Asp39-His69-Ser224).
  • It is stable in a broad range of environments: pH, buffer salts, detergents (SDS), and temperature.
  • In the presence of 0.1-0.5% SDS, proteinase K retains activity and will digest a variety of proteins and nucleases in DNA preparations without compromising the integrity of the isolated DNA.
Specifications

Specifications

39450-01-6
6.2 to 6.8 (20°C, 10 g/L in H2O)
Store at 2°C to 8°C
Endopeptidase K, Proteinase K from Tritirachium album, Tritirachium alkaline proteinase
SDS
Product Certifications