Learn More
Trajan Scientific and Medical™ ProteCol™ Stainless Steel Unions
Precision engineered for capillary LC. Trajan Scientific and Medical ProteCol™ Stainless Steel Unions offer complete versatility for connecting capillary tubing.
Brand:  Trajan Scientific and Medical™ 212008
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15299203
  • 126.00 EUR / Pack of 2
Estimated Shipment: 03-06-2024
Description
  • Leak free connections
  • Replacement parts such as nuts and ferrules
Specifications
Specifications
Union
Size: 1/16 in. to 0.36 mm