missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propargyltriphenylphosphonium bromide, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Propargyltriphenylphosphonium bromide can be used in the alcylation of amines. It is a commonly used as a Wittig reagent.
Solubility
Soluble in methanol.
Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
Propargyltriphenylphosphonium bromide can be used in the alcylation of amines. It is a commonly used as a Wittig reagent.
Solubility
Soluble in methanol.
Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
Specifications
Melting Point | 177°C to 182°C |
Odor | Odorless |
Linear Formula | HC≡CCH2P(C6H5)3Br |
Quantity | 5 g |
Beilstein | 3579775 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in methanol. |
IUPAC Name | triphenyl(prop-2-ynyl)phosphanium;bromide |
Formula Weight | 381.26 |
Percent Purity | 97% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'