Learn More
Invitrogen™ Prolactin (PRL) Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMPRLX5
Additional Details : Weight : 0.33800kg
Description
The Mouse Prolactin (PRL) ELISA quantitates Ms PRL in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms PRL. Principle of the method The Mouse PRL solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Prolactin (PRL) or Luteotropic hormone (LTH) is a peptide hormone discovered by Dr. Henry Friesen, primarily associated with lactation. In breastfeeding, the act of an infant sucking the nipple stimulates the production of prolactin, which fills the breast with milk via a process called lactogenesis, in preparation for the next feed. Oxytocin, another hormone, is also released, which triggers milk let-down.Specifications
P06879 | |
30 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
19109 | |
<12% | |
Assay Diluent Concentrate, Adhesive Plate Covers, Chromogen, Biotinylated Detection Antibody, Pre-coated 96 well plates, Wash Buffer, Stop Solution, SAV-HRP, Standard | |
5 x 96 Tests | |
Endocrinology | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
27.43 pg/mL to 20,000 pg/mL | |
30 pg/mL to 20,000 pg/mL | |
Biotin | |
Hormones & Receptors | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP- 5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, PSelectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF. | |
Mouse | |
Colorimetric Microplate Reader | |
AV290867,Gha11a1,Prl | |
<10% | |
HRP | |
RUO | |
Plasma, 20 μ;L; Serum, 20 μ;L; Supernatant, 50 μ;L | |
RP23-351I6.2, AV2908671a1, Prl | |
1 hr. 20 min. |