Learn More
Techsit™ Pro-Tech Plus Clean Room ESD High Chair
This Cleanroom Class 5 vinyl chair boasts a foot ring on glides. Fully ergonomic, adjustable and enclosed mechanisms.
Brand: Techsit™ TS7911W
Additional Details : Weight : 19.00000kg
Description
- Adjustable foot ring on glides
- ESD/Anti-static vinyl is easy to clean
- Fully adjustable seat height, back height and back rake
- Ergonomically designed and contoured seat and back
- 5 year guarantee includes vinyl
- BS EN61340-5-1 Conductivity: 104 to 106Ω
- Fully enclosed mechanisms
Specifications
Yes | |
Cleanrooms, ESD Environments | |
Yes | |
No | |
Glide | |
Ergonomic | |
500 mm | |
Steel | |
White |
5 Year | |
No | |
Yes | |
15 kg | |
Yes | |
590 to 850 mm | |
Plastic | |
Yes | |
High Bench Height |