missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Primary Color Solution is a ready-to-use coloration standard presented in high-quality, tamper-evident bottles for use as a calibration and/or quality control standard. It is prepared with cobalt chloride, hydrochloric acid and water.
- Manufactured according to European Pharmocopoeia Chapter 2 guidelines with a Balance Certificate of Calibration issued in accordance with ISO 17025
- Prepared gravimetrically on a weight/weight basis using OIML traceable weights
- Concentration verified using a high-performance spectrophotometer calibrated using ISO Guide 34 accredited spectrophotometric standards
- The Primary Color Solutions can be combined in precise ratios to form the Standard Color Solutions
Specifications
Specifications
| Color | Red |
| CAS | 7646-79-9 |
| Molecular Formula | Cl2Co |
| MDL Number | MFCD00010938 |
| Synonym | cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride |
| InChI Key | GVPFVAHMJGGAJG-UHFFFAOYSA-L |
| SMILES | [Cl-].[Cl-].[Co++] |
| IUPAC Name | λ²-cobalt(2+) dichloride |
| Molecular Weight (g/mol) | 129.83 |
| Formula Weight | 129.84g/mol |
| Show More |
Safety and Handling
Product Identifier
- PRIMARY RED SOLUTION
- Danger
- Respiratory sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Acute toxicity Category 4
- Skin sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H410-Very toxic to aquatic life with long lasting effects.
- H317-May cause an allergic skin reaction.
- H302-Harmful if swallowed.
- P284-[In case of inadequate ventilation] wear respiratory protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: cobalt dichloride, hydrochloric acid 37%
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
