Learn More
PREX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553288
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GLGFQIRGFG PSVVHAVGRG TVAAAAGLHP GQCIIKVNGI NVSKETHASV IAHVTACRKY RRPTKQDSIQ WVYNSIESAQ EDLQKSHSKP PGDEAGDAFD CKVEEVIDKF NTMAIIDGKK EHVSLTVDNV HLEYGVVYEY DSTAGIKCNV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
PREX2 Polyclonal antibody specifically detects PREX2 in Human samples. It is validated for Immunohistochemistry.Specifications
PREX2 | |
Polyclonal | |
Unconjugated | |
PREX2 | |
6230420N16Rik; AI316880; AI553603; C030045D06Rik; D430013K02; DEP domain containing 2; DEP domain-containing protein 2; DEP.2; Depdc2; phosphatidylinositol 3,4,5-trisphosphate-dependent Rac exchanger 2 protein; phosphatidylinositol-3,4,5-trisphosphate dependent Rac exchange factor 2; phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 2; PPP1R129; Prex2; P-Rex2; protein phosphatase 1, regulatory subunit 129; ptdIns(3,4,5)-dependent Rac exchanger 2; si:ch211-278g15.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80243 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q70Z35 | |
PREX2 | |
Recombinant protein corresponding to Human PREX2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only