Learn More
IBA Lifesciences™ PPI (2-10) Peptide ALWMRLLPL (HLA-A*0201)
Single peptide synthesized as presented by the MHC Class I allele.
Brand: IBA Lifesciences™ 6-7095-901
Additional Details : Weight : 0.01000kg
Description
PPI (2-10) peptide ALWMRLLPL (HLA-A*0201) is a single peptide for stimulation of T cells. The peptide from preproinsulin (PPI) is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
- Peptides have been purified by HPLC using a water and acetonitrile gradient
- Endotoxin-free
Specifications
>85% | |
T Cell Stimulation | |
1 mg |
Endotoxin-free | |
Yes |