Learn More
Brady™ PowerMark™ Industrial Grade Single Color Labeling Ribbon
Brady's BBP85 and PowerMark™ industrial grade print ribbon is super-tough and smear-proof to ensure your printed text last for years outdoors and in industrial areas.
Brand:  Brady™ 013513
Additional Details : Weight : 0.62370kg
 View more versions of this product
Product Code. 11592605
  • 238.00 EUR / 1 box
Estimated Shipment: 27-05-2024
Description
  • Brady's BBP85 and PowerMark™ industrial grade print ribbon is super-tough and smear-proof to ensure your printed text last for years outdoors and in industrial areas.
Specifications
Specifications
Printer Ribbon
BBP85 Industrial Sign and Label Printer, PowerMark Sign and Label Printer, PowerMark Industrial Sign and Label Printer
6.22 in.
158 mm
BBP85 Ribbon - White Ribbon 158 mm
Printed text last for years outdoors and in industrial areas
White
65.6 yd.
60 m
60 m x 158 mm
Ink side in