missing translation for 'onlineSavingsMsg'
Learn More

Brady™ PowerMark™ Industrial Grade Single Color Labeling Ribbon

Brady's BBP85 and PowerMark™ industrial grade print ribbon is super-tough and smear-proof to ensure your printed text last for years outdoors and in industrial areas.

Brand:  Brady™ 013515

Additional Details : Weight : 0.74840kg

 View more versions of this product

Product Code. 11512615

  • 268.00 EUR / 1 box
Estimated Shipment: 28-05-2024
to see stock.
Add to basket

Description

Description

  • Brady's BBP85 and PowerMark™ industrial grade print ribbon is super-tough and smear-proof to ensure your printed text last for years outdoors and in industrial areas.
Specifications

Specifications

Printer Ribbon
BBP85 Industrial Sign and Label Printer, PowerMark Sign and Label Printer, PowerMark Industrial Sign and Label Printer
8.66 in.
220 mm
BBP85 Ribbon - Blue Ribbon 220 mm
Printed text last for years outdoors and in industrial areas
Blue
65.6 yd.
60 m
60 m x 220 mm
Ink side in
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers