Learn More
Haldenwanger™ Porcelain Desiccator Plates
Use these plates for a variety of applications. Haldenwanger™ Porcelain Desiccator Plates feature a smooth internal finish, constructed from optimized laboratory porcelain.
39.20 EUR - 164.30 EUR
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
10109200
|
Haldenwanger™
119C/190 |
190 mm |
86.87 EUR
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
10653671
|
Haldenwanger™
119C/235 |
235 mm |
132.50 EUR
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
10241481
|
Haldenwanger™
119C/280 |
280 mm |
164.30 EUR
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10557272
|
Haldenwanger™
119C/90 |
90 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10211291
|
Haldenwanger™
119C/140 |
140 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Available in a variety of sizes
- Complies with DIN VDE 0335, type C 100
Specifications
Desiccator Plate | |
Porcelain |