missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinylpyrrolidone, M.W. 10,000, Thermo Scientific Chemicals
26.60 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Polyvinylpyrrolidone is a polymer used as a pharmaceutical aid, complexing agent, and solubilizer. PVP was used as a plasma volume expander for trauma victims. It acts as diluent, solvent and cleaning agent in various kinds of industries. It also acts as an adjuvant, emulsifier, filtering aid, tableting aid, thickener.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Powder |
Merck Index | 14,7697 |
Solubility Information | Soluble in water and most organic solvents. |
Quantity | 25 g |
Formula Weight | 10 kd |
Chemical Name or Material | Polyvinylpyrrolidone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'